Anti-GRK7 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP59819_P050EA 512 CHF
AVIVARP59819_P050
Anti-GRK7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GRK7 (G protein-coupled receptor kinase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR