Anti-HMGN2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP58365_P050EA 512 CHF
AVIVARP58365_P050
Anti-HMGN2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HMGN2 (high mobility group nucleosomal binding domain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR