Anti-SNF8 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP57950_P050EA 512 CHF
AVIVARP57950_P050
Anti-SNF8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SNF8 (SNF8, ESCRT-II complex subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR