Anti-PEX26 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP57067_P050EA 512 CHF
AVIVARP57067_P050
Anti-PEX26 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PEX26 (peroxisomal biogenesis factor 26)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR