Anti-MINA Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP39721_P050 ARP39721_T100
AVIVARP39721_P050EA 533 CHF
AVIVARP39721_P050 AVIVARP39721_T100
Anti-MINA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MINA (MYC induced nuclear antigen)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now