Anti-GTF2F2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP31437_P050 ARP31437_T100
AVIVARP31437_P050EA 533 CHF
AVIVARP31437_P050 AVIVARP31437_T100
Anti-GTF2F2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GTF2F2 (general transcription factor IIF, polypeptide 2, 30kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR