Anion exchange columns, Dionex™, IonPac™ AG24
Lieferant: DIONEX
064151 064153
DION064151EA 0 CHF
DION064151 DION064153
Anion exchange columns, Dionex™, IonPac™ AG24
Chromatographiesäulen
The IonPac™ AS24 is a high capacity hydroxide selective anion exchange column optimised for the separation of haloacetic acids (HAAs) as well as a variety of environmental anions. The capacity and selectivity enables analysis of haloacetic acids in drinking water at low μg/l concentrations. The column is specifically designed for separation of haloacetic acids in drinking water prior to MS or MS/MS detection and is the specified column in US EPA Method 557.
  • Determine HAAs in high-ionic strength matrices without sample pretreatment
  • Column selectivity is
More Product Information
Order Now
TECHNISCHE DATEN
Environmentally Preferable