Washed pooled cells, human blood, non sterile
Lieferant: Rockland Immunochemicals
R407-0050 R407-0100
ROCKR407-0050EA 1510 CHF
ROCKR407-0050 ROCKR407-0100
Washed pooled cells, human blood, non sterile
Blut
Human whole blood is washed to remove the platelet rich plasma, buffy coat layer and leukocytes (WBC). It is used for complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents. Store cells at 4 °C prior to opening.