Examination gloves, PVC, TT® Clear PF
Lieferant: MEDICA EUROPE MAXXIM
480301 480302 480303 480304
MAXX480301EA 0 CHF
MAXX480301 MAXX480302 MAXX480303 MAXX480304
Examination gloves, PVC, TT® Clear PF
Handschuhe
These gloves contain no latex, NRL proteins, or known accelerators. They are therefore suitable for people with a risk of allergy (type I and IV).
  • Non sterile, powder-free, smooth finish
  • Polyurethane (PU) coated which ensures that they are easy to put on
  • Good touch sensitivity
  • High tensile strength
Colour: Natural
EN 455; AQL 1,5
Zertifizierungen: EN 455; AQL 1,5