Bowls, open edge
Lieferant: CONTACTO
1525/150 1525/160 1525/220 1525/240
216-7144EA 34 CHF
216-7144 216-7145 216-7146 216-7147
Bowls, open edge
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Stainless steel, rust-proof, 18/10.
  • Fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, very good thermal resistance
Open edge shallow multipurpose bowl with satin finish.
Weight: 130 g
Order Now
TECHNISCHE DATEN
Environmentally Preferable