Latex gloves, Veniclean 1340
Lieferant: DELTA PLUS
V1340/6-7 V1340/7-8 V1340/8-9
DELPV1340/6-7EA 22.2 CHF
DELPV1340/6-7 DELPV1340/7-8 DELPV1340/8-9
Latex gloves, Veniclean 1340
Handschuhe
These disposable, waterproof gloves provide limited protection against chemical splashes.
  • Non sterile, powder-free, smooth finish
  • High dexterity: Suitable for precision handling
  • Compatible with all types of foods
  • Ambidextrous
Ideal for use in laboratories, hospitals, beauty salons, care facilities and in the pharma industry.
Length: 240 mm
Thickness: 0,125 mm
Colour: Natural
EN420, EN374, AQL 1.5
Zertifizierungen: EN 455-1/2/3, EN ISO 374-1 Type C (KT), EN ISO 374-5 VIRUS; AQL 1,5
Food contact approved