Volumetric flasks, class A, blue markings
Lieferant: NEUBERT VOLUME GLASSWAERE
0294-50-0025 0294-50-0250 0294-10-1000 0294-10-0150 0294-10-0250 0294-50-0300 0294-50-0150 0294-10-0100 0294-50-0901 0294-10-0300 0294-10-0400 0294-10-600034 0294-10-0030 0294-10-0151 0294-50-0151 0294-50-0302 0294-50-5000 0294-50-2000 0294-50-300029 0294-10-0001 0294-10-0002
NEUB0294-50-0025EA 102 CHF
NEUB0294-50-0025 NEUB0294-50-0250 NEUB0294-10-1000 NEUB0294-10-0150 NEUB0294-10-0250 NEUB0294-50-0300 NEUB0294-50-0150 NEUB0294-10-0100 NEUB0294-50-0901 NEUB0294-10-0300 NEUB0294-10-0400 NEUB0294-10-600034 NEUB0294-10-0030 NEUB0294-10-0151 NEUB0294-50-0151 NEUB0294-50-0302 NEUB0294-50-5000 NEUB0294-50-2000 NEUB0294-50-300029 NEUB0294-10-0001 NEUB0294-10-0002
Volumetric flasks, class A, blue markings
Kolben Messkolben
Borosilicate glass, grade 3.3, with glass stopper.
  • Conformity certified with dated batch identification
  • With batch certificate
  • With one graduation mark