Plier wrench
Lieferant: Knipex
238-5515EA 254 CHF
238-5515
Plier wrench
Schraubenschlüssel
The plier wrench combines the jobs of a gripping plier and a wrench in a single tool with flexible adjustment of all widths up to the specified maximum size. Ideal for for gripping, holding, pressing and bending workpieces.
  • Adjustable tightening tool
  • Parallel jaws give a more solid grip
  • Zero backlash jaw pressure prevents damage to edges
  • Easily adjusted at the touch of a button on the workpiece
  • Smooth jaws for damage free installation of plated fittings