GC septa, Marathon
Lieferant: Thermo Fisher Scientific
313P3211 313P3233 313P3240
555-8180EA 185 CHF
555-8180 555-8182 555-8184
GC septa, Marathon
Septen
Made of low-bleed silicone, these septa have excellent mechanical properties and are ideal for demanding GC-MS applications.
  • Pre-pierced for reliable performance
  • Op to 400 injections per septa
  • Maximum temperature 400 °C