Cation exchange HPLC columns, Dionex™, IonPac™ CS14
Lieferant: DIONEX
044123 044124
DION044123EA 0 CHF
DION044123 DION044124
Cation exchange HPLC columns, Dionex™, IonPac™ CS14
Chromatographiesäulen
CS14 columns are used for the determination of aliphatic amines, aromatic amines and polyamines by using methane sulfonic acid or sulfuric acid eluents.
  • Compatible with organic solvents
  • For moderately hydrophobic amines
Used for the sample matrices include environmental water, power plant waters treated with ammonium, morpholine, ethanolamine, and also used for industrial solvents.