Diatomaceous earth sorbents, ISOLUTE HM-N
Lieferant: BIOTAGE
9800-1000
9800-5000
BITA9800-1000EA
201.73
CHF
BITA9800-1000
BITA9800-5000
Diatomaceous earth sorbents, ISOLUTE HM-N
Chromatographie-Sorbentien
SPE-Sorbentien
ISOLUTE HM-N, a modified form of diatomaceous earth. The material is used for applications requiring an inert support as part of the sample preparation process (e.g. SFE).