Diatomaceous earth sorbents, ISOLUTE HM-N
Lieferant: BIOTAGE
9800-1000 9800-5000
BITA9800-1000EA 201.73 CHF
BITA9800-1000 BITA9800-5000
Diatomaceous earth sorbents, ISOLUTE HM-N
Chromatographie-Sorbentien SPE-Sorbentien
ISOLUTE HM-N, a modified form of diatomaceous earth. The material is used for applications requiring an inert support as part of the sample preparation process (e.g. SFE).