Anion exchange HPLC columns, Dionex™, CarboPac™ MA1
Lieferant: DIONEX
044066 044067
DION044066EA 0 CHF
DION044066 DION044067
Anion exchange HPLC columns, Dionex™, CarboPac™ MA1
Chromatographiesäulen
This column can be used for sensitive, rugged and reliable separations of reduced sugars without the need for derivatisation. This high capacity, strong anion exchange column efficiently separates reduced monosaccharides and disaccharides commonly found in food products and physiological samples.