Isolierung, Thinsulate™
Lieferant: 3M
P150 P100
MMMAP150EA 596 CHF
MMMAP150 MMMAP100
Isolierung, Thinsulate™
Isolierung
This insulation fabric can be used with outdoor clothing (jackets and trousers). It is designed to keep the body warm in cold weather conditions.
Type P is a mid-loft, soft insulation fabric with excellent product features, such as washability up to 60 °C. The insulation is also suitable for dry cleaning.
Some types are suitable for extreme weather conditions. Cold temperatures may require multiple layers of the insulation fabric.