5-Brom-2-methoxy-3-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS35357G1 AS35357G10 AS35357G5
ACTIAS35357G1EA 0 CHF
ACTIAS35357G1 ACTIAS35357G10 ACTIAS35357G5
5-Brom-2-methoxy-3-methylpyridin
5-Brom-2-methoxy-3-methylpyridin
Formel: C₇H₈BrNO
MDL Number: MFCD07375124
CAS-Nummer: 760207-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR