Ethyl-4-piperidincarboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl piperidine-4-carboxylate, Ethyl isonipecotate, Ethyl 4-Piperidinecarboxylate

AS34462G10 AS34462G100
ACTIAS34462G10EA 0 CHF
ACTIAS34462G10 ACTIAS34462G100
Ethyl-4-piperidincarboxylat
Ethyl-4-piperidincarboxylat
Formel: C₈H₁₅NO₂
MW: 157.21 g/mol
MDL Number: MFCD00006003
CAS-Nummer: 1126-09-6
EINECS: 214-416-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR