(6-Chlor-3-pyridinyl)methanol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
6-Chloropyridine-3-methanol, 2-Chloro-5-hydroxymethylpyridine, 6-Chloro-3-(hydroxymethyl)pyridine
AS20766G25
AS20766G5
ACTIAS20766G25EA
0
CHF
ACTIAS20766G25
ACTIAS20766G5
(6-Chlor-3-pyridinyl)methanol
(6-Chlor-3-pyridinyl)methanol
Formel:
C₆H₆ClNO MW: 143,57 g/mol |
MDL Number:
MFCD01075226 CAS-Nummer: 21543-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...