2-Fluor-6-formylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Fluoropicolinaldehyde, 6-Fluoropyridine-2-carboxaldehyde

AS31202G1 AS31202G5
ACTIAS31202G1EA 0 CHF
ACTIAS31202G1 ACTIAS31202G5
2-Fluor-6-formylpyridin
2-Fluor-6-formylpyridin
Formel: C₆H₄FNO
MW: 125,1 g/mol
MDL Number: MFCD03095149
CAS-Nummer: 208110-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR