2-Formyl-6-methoxypyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Methoxypicolinaldehyde, 6-Methoxypyridine-2-carboxaldehyde, 6-Methoxypyridine-2-carbaldehyde

AS23921G1 AS23921G25 AS23921G5
ACTIAS23921G1EA 0 CHF
ACTIAS23921G1 ACTIAS23921G25 ACTIAS23921G5
2-Formyl-6-methoxypyridin
2-Formyl-6-methoxypyridin
Formel: C₇H₇NO₂
MDL Number: MFCD05664030
CAS-Nummer: 54221-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR