4'-(Trifluormethyl)biphenyl-4-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid

AS30283G1 AS30283G5
ACTIAS30283G1EA 0 CHF
ACTIAS30283G1 ACTIAS30283G5
4'-(Trifluormethyl)biphenyl-4-carbonsäure
4'-(Trifluormethyl)biphenyl-4-carbonsäure
Formel: C₁₄H₉F₃O₂
MDL Number: MFCD03424606
CAS-Nummer: 195457-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR