4'-(Trifluormethyl)biphenyl-3-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS30293G1 AS30293G5
ACTIAS30293G1EA 0 CHF
ACTIAS30293G1 ACTIAS30293G5
4'-(Trifluormethyl)biphenyl-3-carbonsäure
4'-(Trifluormethyl)biphenyl-3-carbonsäure
Formel: C₁₄H₉F₃O₂
MW: 266,22 g/mol
MDL Number: MFCD03424604
CAS-Nummer: 199528-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR