4'-Brom-2'-hydroxyacetophenon
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
1-(4-Bromo-2-hydroxyphenyl)ethanone, 1-(4-Bromo-2-hydroxyphenyl)ethan-1-one
AS1815G1
AS1815G5
ACTIAS1815G1EA
0
CHF
ACTIAS1815G1
ACTIAS1815G5
4'-Brom-2'-hydroxyacetophenon
4'-Brom-2'-hydroxyacetophenon
Formel:
C₈H₇BrO₂ MW: 215,05 g/mol |
MDL Number:
MFCD03428533 CAS-Nummer: 30186-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...