3-Fluor-4-(trifluormethyl)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS34947G1 AS34947G10 AS34947G5
ACTIAS34947G1EA 0 CHF
ACTIAS34947G1 ACTIAS34947G10 ACTIAS34947G5
3-Fluor-4-(trifluormethyl)benzonitril
3-Fluor-4-(trifluormethyl)benzonitril
Formel: C₈H₃F₄N
Siedepunkt: 212 °C (1013 hPa)
Melting Pt: 45…48 °C
MDL Number: MFCD00236280
CAS-Nummer: 231953-38-1
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR