2-Fluor-5-(trifluormethyl)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS40242G10 AS40242G100
ACTIAS40242G10EA 0 CHF
ACTIAS40242G10 ACTIAS40242G100
2-Fluor-5-(trifluormethyl)benzonitril
Formel: C₈H₃F₄N
Siedepunkt: 99 °C (1013 hPa)
MDL Number: MFCD00061280
CAS-Nummer: 4088-84-0
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR