3-Fluor-2-pyridincarbonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Fluoro-2-pyridinecarbonitrile, 2-Cyano-3-fluoropyridine, 3-Fluoropicolinonitrile

AS34900G1 AS34900G5
ACTIAS34900G1EA 0 CHF
ACTIAS34900G1 ACTIAS34900G5
3-Fluor-2-pyridincarbonitril
3-Fluor-2-pyridincarbonitril
Formel: C₆H₃FN₂
MW: 122,1 g/mol
MDL Number: MFCD06797501
CAS-Nummer: 97509-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR