α,α,α-Trifluor-m-xylol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Methyl-3-(trifluoromethyl)benzene, 3-(Trifluoromethyl)toulene, 3-Methylbenzotrifluoride

AS34443G1 AS34443G10 AS34443G5
ACTIAS34443G1EA 0 CHF
ACTIAS34443G1 ACTIAS34443G10 ACTIAS34443G5
α,α,α-Trifluor-m-xylol
α,α,α-Trifluor-m-xylol
Formel: C₈H₇F₃
MW: 160.14 g/mol
Siedepunkt: 127 °C (1013 hPa)
Dichte: 1.148 g/cm³ (20 °C)
MDL Number: MFCD00190105
CAS-Nummer: 401-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR