Rifampicin
Lieferant: Cayman Chemical
Synonyms: Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP
14423-100 14423-500 14423-1 14423-5
CAYM14423-100EA 65.5 CHF
CAYM14423-100 CAYM14423-500 CAYM14423-1 CAYM14423-5
Rifampicin
Rifampicin
Formel: C₄₃H₅₈N₄O₁₂
Melting Pt: 183…188 °C
MDL Number: MFCD00151389
CAS-Nummer: 13292-46-1
EINECS: 236-312-0
Merck Index: 12,08382