2,2'-Dibrombiphenyl (PBB Nr. 4)

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Bromo-2-(2-bromophenyl)benzene, 2,2'-Dibromobiphenyl

AS33298G1 AS33298G10 AS33298G5
ACTIAS33298G1EA 0 CHF
ACTIAS33298G1 ACTIAS33298G10 ACTIAS33298G5
2,2'-Dibrombiphenyl (PBB Nr. 4)
2,2'-Dibrombiphenyl (PBB Nr. 4)
Formel: C₁₂H₈Br₂
MW: 312 g/mol
MDL Number: MFCD00093707
CAS-Nummer: 13029-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR