Picolinoylchlorid Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Pyridine-2-carbonyl chloride hydrochloride

AS33309G1 AS33309G5
ACTIAS33309G1EA 0 CHF
ACTIAS33309G1 ACTIAS33309G5
Picolinoylchlorid Hydrochlorid
Picolinoylchlorid Hydrochlorid
Formel: C₆H₅Cl₂NO
MW: 178,0160 g/mol
MDL Number: MFCD00060193
CAS-Nummer: 39901-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR