(R)-(-)-Ethyl-3-piperidincarboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl (R)-(-)-3-Piperidinecarboxylate, (R)-Ethyl piperidine-3-carboxylate

AS32729G1 AS32729G10 AS32729G5
ACTIAS32729G1EA 0 CHF
ACTIAS32729G1 ACTIAS32729G10 ACTIAS32729G5
(R)-(-)-Ethyl-3-piperidincarboxylat
(R)-(-)-Ethyl-3-piperidincarboxylat
Formel: C₈H₁₅NO₂
MW: 157.2102 g/mol
MDL Number: MFCD03093637
CAS-Nummer: 25137-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR