5-Brom-2-hydroxy-3-picolin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Bromo-3-methylpyridin-2-ol, 5-Bromo-2-hydroxy-3-methylpyridine, 5-Bromo-3-methylpyridin-2(1H)-one

AS22545G25 AS22545G5
ACTIAS22545G25EA 57 CHF
ACTIAS22545G25 ACTIAS22545G5
5-Brom-2-hydroxy-3-picolin
5-Brom-2-hydroxy-3-picolin
Formel: C₆H₆BrNO
MW: 188,02 g/mol
MDL Number: MFCD03427657
CAS-Nummer: 89488-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR