4-Iod-1-tritylimidazol

Lieferant: ACTIVATE SCIENTIFIC

AS50087G1 AS50087G25 AS50087G5
ACTIAS50087G1EA 0 CHF
ACTIAS50087G1 ACTIAS50087G25 ACTIAS50087G5
4-Iod-1-tritylimidazol
4-Iod-1-tritylimidazol
Formel: C₂₂H₁₇IN₂
MDL Number: MFCD02179542
CAS-Nummer: 96797-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR