4-Brom-1-tritylimidazol

Lieferant: ACTIVATE SCIENTIFIC

AS51147G1 AS51147G5
ACTIAS51147G1EA 0 CHF
ACTIAS51147G1 ACTIAS51147G5
4-Brom-1-tritylimidazol
4-Brom-1-tritylimidazol
Formel: C₂₂H₁₇BrN₂
MDL Number: MFCD00185657
CAS-Nummer: 87941-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR