6-(Trifluormethyl)picolinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-(Trifluoromethyl)-2-pyridinecarboxylic acid, 6-(Trifluoromethyl)picolinic acid, 2-(Trifluoromethyl)-6-pyridinecarboxylic acid, 6-(Trifluoromethyl)pyridine-2-carboxylic acid

AS31886G1 AS31886G5
ACTIAS31886G1EA 0 CHF
ACTIAS31886G1 ACTIAS31886G5
6-(Trifluormethyl)picolinsäure
6-(Trifluormethyl)picolinsäure
Formel: C₇H₄F₃NO₂
MW: 191,11 g/mol
MDL Number: MFCD02180819
CAS-Nummer: 131747-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR