2-Brom-6-(brommethyl)pyridin

Lieferant: ACTIVATE SCIENTIFIC

AS36394G1 AS36394G5
ACTIAS36394G1EA 0 CHF
ACTIAS36394G1 ACTIAS36394G5
2-Brom-6-(brommethyl)pyridin
2-Bromo-6-(bromomethyl)pyridine
Formel: C₆H₅Br₂N
MW: 250,9200 g/mol
MDL Number: MFCD07368202
CAS-Nummer: 83004-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR