3,5-Dichlor-4-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33432G1 AS33432G5
ACTIAS33432G1EA 0 CHF
ACTIAS33432G1 ACTIAS33432G5
3,5-Dichlor-4-nitropyridin
3,5-Dichloro-4-nitropyridine
Formel: C₅H₂Cl₂N₂O₂
MW: 192,99 g/mol
MDL Number: MFCD06254584
CAS-Nummer: 433294-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR