3,5-Dichlor-4-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33451G1 AS33451G5
ACTIAS33451G1EA 0 CHF
ACTIAS33451G1 ACTIAS33451G5
3,5-Dichlor-4-fluorpyridin
3,5-Dichloro-4-fluoropyridine
Formel: C₅H₂Cl₂FN
MDL Number: MFCD07368620
CAS-Nummer: 916791-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR