3,5-Dibrom-2-chlorpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Chloro-3,5-dibromopyridine

AS4035G100 AS4035G5
ACTIAS4035G100EA 0 CHF
ACTIAS4035G100 ACTIAS4035G5
3,5-Dibrom-2-chlorpyridin
3,5-Dibrom-2-chlorpyridin
Formel: C₅H₂Br₂ClN
MW: 271.34 g/mol
MDL Number: MFCD00233991
CAS-Nummer: 40360-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR