6-Nitropicolinsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Nitropyridine-2-carboxylic acid

AS1126G1 AS1126G5
ACTIAS1126G1EA 0 CHF
ACTIAS1126G1 ACTIAS1126G5
6-Nitropicolinsäure
6-Nitropicolinsäure
Formel: C₆H₄N₂O₄
MW: 168,11 g/mol
MDL Number: MFCD04114205
CAS-Nummer: 26893-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR