4-Nitropicolinsäure

Lieferant: ACTIVATE SCIENTIFIC

AS1164G1 AS1164G5
ACTIAS1164G1EA 0 CHF
ACTIAS1164G1 ACTIAS1164G5
4-Nitropicolinsäure
4-Nitropicolinsäure
Formel: C₆H₄N₂O₄
MW: 168,110 g/mol
MDL Number: MFCD00191562
CAS-Nummer: 13509-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR