4-Brom-2-nitrobenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32031G1 AS32031G5
ACTIAS32031G1EA 0 CHF
ACTIAS32031G1 ACTIAS32031G5
4-Brom-2-nitrobenzonitril
4-Brom-2-nitrobenzonitril
Formel: C₇H₃BrN₂O₂
MDL Number: MFCD00087225
CAS-Nummer: 79603-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR