8-Amino-2-methylchinolin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-Methylquinolin-8-amine, 8-Aminoquinaldine
AS31716G1
AS31716G5
ACTIAS31716G1EA
0
CHF
ACTIAS31716G1
ACTIAS31716G5
8-Amino-2-methylchinolin
8-Amino-2-methylquinoline
Formel:
C₁₀H₁₀N₂ MW: 158.1998 g/mol |
MDL Number:
MFCD00023998 CAS-Nummer: 18978-78-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...