2-Fluor-m-kresol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-3-methylphenol

AS35083G1 AS35083G10 AS35083G5
ACTIAS35083G1EA 0 CHF
ACTIAS35083G1 ACTIAS35083G10 ACTIAS35083G5
2-Fluor-m-kresol
2-Fluor-m-kresol
Formel: C₇H₇FO
MW: 126,1300 g/mol
MDL Number: MFCD09835186
CAS-Nummer: 77772-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR