4-Methoxy-2-methylbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Methyl-4-methoxybenzaldehyde

AS33836G1 AS33836G5
ACTIAS33836G1EA 0 CHF
ACTIAS33836G1 ACTIAS33836G5
4-Methoxy-2-methylbenzaldehyd
4-Methoxy-2-methylbenzaldehyde
Formel: C₉H₁₀O₂
MDL Number: MFCD02261771
CAS-Nummer: 52289-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR