2-Methoxy-4-methylbenzaldehyd

Lieferant: ACTIVATE SCIENTIFIC

AS23308G1 AS23308G5
ACTIAS23308G1EA 68 CHF
ACTIAS23308G1 ACTIAS23308G5
2-Methoxy-4-methylbenzaldehyd
2-Methoxy-4-methylbenzaldehyde
Formel: C₉H₁₀O₂
MW: 150,1700 g/mol
MDL Number: MFCD06248007
CAS-Nummer: 57415-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR